missing translation for 'onlineSavingsMsg'
Learn More

USF1 Antibody (2A7), Novus Biologicals™

Product Code. 18364088 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364088 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364088 Supplier Novus Biologicals Supplier No. H00007391M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

USF1 Monoclonal antibody specifically detects USF1 in Human samples. It is validated for Western Blot, ELISA, Dot Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen USF1
Applications Western Blot, ELISA, Dot Blot, Sandwich ELISA
Classification Monoclonal
Clone 2A7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_009053
Gene Alias BHLHB11, bHLHb11FCHL1, Class B basic helix-loop-helix protein 11, FCHL, HYPLIP1, Major late transcription factor 1, MLTF, MLTFI, UEF, upstream stimulatory factor 1, upstream transcription factor 1, USF
Host Species Mouse
Immunogen USF1 (NP_009053, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MKGQQKTAETEEGTVQIQEGAVATGEDPTSVAIASIQSAATFPDPNVKYVFRTENGGQVMYRVIQVSEGQLDGQTEGTGAISGYPATQSMTQAVIQGAFTSDDAVDTEGT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7391
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.