missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USE1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25218-1-AP-150UL
This item is not returnable.
View return policy
Description
MDS032 belongs to the USE1 family and is a component of a SNARE complex which may be involved in targeting and fusion of Golgi-derived retrograde transport vesicles with the ER.Specifications
| USE1 | |
| Polyclonal | |
| Unconjugated | |
| USE1 | |
| MDS032, P31, SLT1, USE1, USE1 like protein, USE1L, Vesicle transport protein USE1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55850 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NZ43 | |
| USE1 | |
| USE1 Fusion Protein Ag18587 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction