missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ USE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112263
This item is not returnable.
View return policy
Description
USE1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
MDS032 belongs to the USE1 family and is a component of a SNARE complex which may be involved in targeting and fusion of Golgi-derived retrograde transport vesicles with the ER.
Specifications
| USE1 | |
| Polyclonal | |
| Unconjugated | |
| Use1 | |
| 2010315L10Rik; 5730403H22Rik; AV002165; D12; Ed2; embryonic development factor 2; fj43b06; hypothetical protein MGC56402; MDS032; P31; Protein D12; protein p31; putative MAPK activating protein PM26; putative MAPK-activating protein PM26; Q-snare; RGD1306660; SLT1; SNARE-like tail-anchored protein 1 homolog; unconventional SNARE in the ER 1; unconventional SNARE in the ER 1 homolog; unconventional SNARE in the ER 1 homolog (S. cerevisiae); USE1; Use1l; USE1-like protein; Vesicle transport protein USE1; wu:fj43b06; zgc:56402 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 55850 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9NZ43 | |
| Use1 | |
| E. coli-derived Human USE1 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction