missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ USE1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17161354 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17161354 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17161354 Supplier Invitrogen™ Supplier No. PA5113496

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USE1 Polyclonal Antibody for Western Blot, ICC/IF, IP, ELISA

MDS032 belongs to the USE1 family and is a component of a SNARE complex which may be involved in targeting and fusion of Golgi-derived retrograde transport vesicles with the ER.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USE1
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Use1
Gene Accession No. Q9CQ56, Q9NZ43
Gene Alias 2010315L10Rik; 5730403H22Rik; AV002165; D12; Ed2; embryonic development factor 2; fj43b06; hypothetical protein MGC56402; MDS032; P31; Protein D12; protein p31; putative MAPK activating protein PM26; putative MAPK-activating protein PM26; Q-snare; RGD1306660; SLT1; SNARE-like tail-anchored protein 1 homolog; unconventional SNARE in the ER 1; unconventional SNARE in the ER 1 homolog; unconventional SNARE in the ER 1 homolog (S. cerevisiae); USE1; Use1l; USE1-like protein; Vesicle transport protein USE1; wu:fj43b06; zgc:56402
Gene Symbols Use1
Host Species Rabbit
Immunogen Recombinant Human Vesicle transport protein USE1 protein (1-146AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55850, 67023
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.