Learn More
IKA USB WD
Product Code. 31910315 Shop All IKA Products
Change view
Click to view available options
Interface:
USB-C, Bluetooth
Unit Size:
Each
Product Code. Interface Unit Size
31910315 USB-C, Bluetooth Each
1 options
Product Code. 31910315 Supplier IKA Supplier No. 0020123620
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
Extends the RCT basic with wireless communication capabilities by simply plugging it in for modern connected laboratory applications.
  • The USB WD USB dongle extends the RCT basic with wireless communication capabilities simply by plugging it in, creating the basis for modern, connected laboratory applications.
  • Enables wireless connectivity to the IKA wireless sensors Pt.Wireless and ETS-D7, as well as to the IKA App.
  • Allows temperature and pH values to be captured wirelessly and displayed in real time.
  • In combination with IKA labworldsoft™, measured values can be documented digitally.
  • Supports wireless device-to-device communication.
  • Simply plug in, connect wirelessly, and expand functionality.
TRUSTED_SUSTAINABILITY
Specifications
Type USB Dongle
Connection Type Black
For Use With (Equipment) IKA RCT Basic 0025008259, RCT Basic Anodized 0020133785
Interface USB-C, Bluetooth
Dimensions (D x W x H) 38.5 x 18 x 10 mm
Voltage 5 V