missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ USB User-ID Stick (with User-ID Licence) for UN75plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN75plus Oven
Unit Size:
Each
Description
- Oven-linked authorization licence (User-ID-program) on Memory-stick
- Prevents undesired manipulation by unauthorised third parties
- When reordering please specify serial number (only for units with TwinDISPLAY)
Specifications
Specifications
| For Use With (Equipment) | UN75plus Oven |
| Type | USB Stick |
| Includes | User ID Licence |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction