Learn More
Applied Biosystems™ USB Sequenase Reaction Buffer
Product Code. 15840783
Change view
Click to view available options
For Use With (Application):
DNA Sequencing
Unit Size:
1mL
Product Code. For Use With (Application) unitSize
15840783 DNA Sequencing 1mL
1 options
Product Code. 15840783 Supplier Applied Biosystems™ Supplier No. 707021ML
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The USB Sequenase Reaction Buffer (5X) is a component of the Sequenase Quick Denature Plasmid DNA Sequencing Kit. This buffer can also be purchased separately.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) DNA Sequencing
Type USB Sequenase Reaction Buffer