Learn More
ATP™ Usb Endoscope with Semi-Rigid Gooseneck Probe
Product Code. 12817875
Encompass_Preferred
Product Code. 12817875
Brand: ATP™ ENDSUSB1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
waterproof camera head, 2m USB connection cable powered by USB PC/laptop connection port probe 880mm x 8.5mm (l x dia.) 60mm to infinity, 640 x 480 pixels ATP