missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ USB™ CycleSeq™ Thermostable DNA Polymerase

Product Code. 15879916
Click to view available options
Quantity:
1000 U
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15879916

Brand: Applied Biosystems™ 792001000UN

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

CycleSeq Thermostable DNA Polymerase 1,000 units, 1mL CycleSeq Dilution Buffer, 1mL 10X CycleSeq Reaction Buffer

USB™ CycleSeq™ Thermostable DNA Polymerase is a thermostable, DNA polymerase that is exonuclease-free and incorporates ddNTPs with a marked level of efficiency over standard thermostable DNA polymerases.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage CycleSeq Thermostable DNA Polymerase 1,000 units
• 1 mL CycleSeq Dilution Buffer
• 1 mL 10X CycleSeq Reaction Buffer

For Use With (Application) DNA Sequencing
Polymerase Thermostable DNA Polymerase
Quantity 1000 U

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.