Learn More
Edmund Buehler™ USB 2.0 interface
Click to view available options
:
USB interface
Description
- Socket type B
- Plug-and-play connection with standard terminal programs
- Complete remote control of the shaker
Specifications
Specifications
| Product Type | USB Interface |
| For Use With (Equipment) | KS 15 A Control, KS 15 B Control, SM 30 A Control, SM 30 B Control, SM 30 C Control, SM 30 AT Control, VKS 75 A Control, VKS 75 B Control |