missing translation for 'onlineSavingsMsg'
Learn More
Learn More
US EPA 23 Metals Standard: Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Tl, V, Zn @ 100 μg/mL in 5% HNO3, tr. Tartaric Acid, tr. HF, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
US EPA 23 Metals Standard: Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Tl, V, Zn @ 100 μg/mL in 5% HNO3, tr. Tartaric Acid, tr. HF
Specifications
Specifications
| Chemical Name or Material | Aluminium/Antimony/Arsenic/Barium/Beryllium/Cadmium/Calcium/Chromium/Cobalt/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Potassium/Selenium/Silver/Sodium/Thallium/Vanadium/Zinc |
| CAS | 7429-90-5, 7440-36-0, 7440-38-2, 7440-39-3, 7440-41-7, 7440-43-9 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Aluminium/Antimony/Arsenic/Barium/Beryllium/Cadmium/Calcium/Chromium/Cobalt/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Potassium/Selenium/Silver/Sodium/Thallium/Vanadium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 100 μg/mL/Antimony 100 μg/mL/Arsenic 100 μg/mL/Barium 100 μg/mL/Beryllium 100 μg/mL/Cadmium 100 μg/mL/Calcium 100 μg/mL/Chromium 100 μg/mL/Cobalt 100 μg/mL/Copper 100 μg/mL/Iron 100 μg/mL/Lead 100 μg/mL/Magnesium 100 μg/mL/Manganese 100 μg/mL/Molybdenum 100 μg/mL/Nickel 100 μg/mL/Potassium 100 μg/mL/Selenium 100 μg/mL/Silver 100 μg/mL/Sodium 100 μg/mL/Thallium 100 μg/mL/Vanadium 100 μg/mL/Zinc 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction