missing translation for 'onlineSavingsMsg'
Learn More

Ursodeoxycholic Acid, MP Biomedicals™

Product Code. 17821431 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17821431 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17821431 Supplier MP Biomedicals Supplier No. 0215825201
  • ONLINE EXCLUSIVE PRICE
    £40.55
    £38.01 / 1g
    Save £2.54
    6% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used extensively to mitigate hepatic and biliary diseases. Ursodeoxycholic Acid, MP Biomedicals is a cell protectant. UDCS may be used to study specific activities ranging from reduction of cholesterol absorption and cholesterol gallstone dissolution to s

Specifications

CAS 128-13-2
Melting Point 203°C to 204°C (literature)
Molecular Formula C24H40O4
MDL Number MFCD00003680
Quantity 1 g
Synonym ursodeoxycholic acid, ursodiol, actigall, ursofalk, ursolvan, delursan, ursodeoxycholate, urso, urso forte, ursodesoxycholic acid
Solubility Information Soluble in Ethanol (50mg/mL, Clear to Very Slightly Hazy).
InChI Key RUDATBOHQWOJDD-UZVSRGJWSA-N
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])[C@@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C
IUPAC Name (4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molecular Weight (g/mol) 392.58
PubChem CID 31401
ChEBI CHEBI:9907
Formula Weight 392.6
Percent Purity ≥99%
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • Ursodeoxycholic Acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ursodeoxycholic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.