missing translation for 'onlineSavingsMsg'
Learn More

Ursodeoxycholic Acid, Mikromol™

Product Code. 17910416
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17910416 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17910416 Supplier Mikromol Supplier No. MM0836.000250
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ursodeoxycholic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ursodeoxycholic Acid
CAS 128-13-2
Analyte or Component Names Ursodeoxycholic Acid
Shipping Condition Room Temperature
Molecular Formula C24 H40 O4
Certifications/Compliance ISO 17025
Synonym Ursodeoxycholic acid, 3α,7β-Dihydroxy-5β-cholan-24-oic acid, 5β-Cholan-24-oic acid, 3α,7β-dihydroxy- (8CI), 5β-Cholanic acid, 3α,7β-dihydroxy- (7CI), 17β-(1-Methyl-3-carboxypropyl)etiocholane-3α,7β-diol, 3α,7β-Dihydroxy-5β-cholan-24-oate, 3α,7β-Dihydroxy-5β-cholan-24-oic acid, 3α,7β-Dihydroxy-5β-cholanic acid, 3α,7β-Dihydroxy-5β-cholanoic acid, 3α,7β-Dihydroxycholanic acid, 7β-Hydroxylithocholic acid, Actigall, Adursal, Arsacol, Cholit-Ursan, Delursan, Desocol, Desol, Destolit, Deursil, Litursol, Lyeton, NSC 683769, Paptarom, Solutrat, UDCA, Urdes, Ursacol, Ursic acid, Urso, Ursobilin, Ursochol, Ursocholic acid, deoxy-, Ursocol, Ursodamor, Ursodeoxycholic acid, Ursodesoxycholic acid, Ursodiol, Ursofalk, Ursolvan, Ursosan, Chenodeoxycholic Acid Impurity A
InChI Formula InChI=1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20+,22+,23+,24-/m1/s1
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name (4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molecular Weight (g/mol) 392.57
Quantity 250 mg
Formula Weight 392.2927 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.