missing translation for 'onlineSavingsMsg'
Learn More

Ursodeoxycholic Acid Methyl Ester, TRC

Product Code. 30450948
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450948 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450948 Supplier Toronto Research Chemicals Supplier No. TRCU8500505G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ursodeoxycholic-1-methyl Ester
CAS 10538-55-3
Purity Grade Notes HPLC
Molecular Formula C25 H42 O4
Quantity 5 g
Synonym (3α,5β,7β)-3,7-Dihydroxycholan-24-oic acid methyl ester, 3α,7β-Dihydroxy-5β-cholan-24-oic acid methyl ester), 3α,7β-Dihydroxy-5β-cholanic acid methyl ester, Methyl 3α,7β-dihydroxy-5β-cholanoate, Methyl ursodeoxycholate, Ursodeoxycholic acid methyl ester
InChI Formula InChI=1S/C25H42O4/c1-15(5-8-22(28)29-4)18-6-7-19-23-20(10-12-25(18,19)3)24(2)11-9-17(26)13-16(24)14-21(23)27/h15-21,23,26-27H,5-14H2,1-4H3/t15-,16+,17-,18-,19+,20+,21+,23+,24+,25-/m1/s1
SMILES COC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name methyl (4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
Molecular Weight (g/mol) 406.6
Formula Weight 406.3083
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.