missing translation for 'onlineSavingsMsg'
Learn More

Ursodeoxycholic acid, MedChemExpress

Product Code. p-200096579
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448507 5 g 5g
16411389 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448507 Supplier Medchem Express Supplier No. HY13771

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ursodeoxycholic acid (Ursodeoxycholate) is a secondary bile acid issued from the transformation of (cheno)deoxycholic acid by intestinal bacteria, acting as a key regulator of the intestinal barrier integrity and essential for lipid metabolism.

Ursodeoxycholic acid acts as signaling molecule, exerting its effects by interacting with bile acid activated receptors, including G-protein coupled bile acid receptor 5 (TGR5, GPCR19) and the farnesoid X receptor (FXR). Ursodeoxycholic acid can be used for the research of a variety of hepatic and gastrointestinal diseases. Orally active.

Specifications

Chemical Name or Material Ursodeoxycholic acid
CAS 128-13-2
Color White
Purity Grade Notes Research
Quantity 5 g
Synonym Ursodeoxycholate Ursodiol UDCA
Solubility Information DMSO : ≥ 100 mg/mL (254.73 mM) ∣H2O : 1 mg/mL (2.55 mM; ultrasonic and warming and heat to 80°C)
SMILES C[C@H](CCC(O)=O)[C@H]1CC[C@@]2([H])[C@]3([H])[C@@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C
Molecular Weight (g/mol) 392.57
Formula Weight 392.57
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.