missing translation for 'onlineSavingsMsg'
Learn More

Ursocholic Acid, TRC

Product Code. 30010436
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010436 10 mg 10mg
30008610 25 mg 25mg
30005189 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010436 Supplier Toronto Research Chemicals Supplier No. TRCU84990010MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2955-27-3
Molecular Formula C24 H40 O5
Molecular Weight (g/mol) 408.57
Synonym (3alpha,5beta,7beta,12alpha)-3,7,12-Trihydroxycholan-24-oic acid, 3alpha,7beta,12alpha-Trihydroxy-5beta-cholan-24-oic acid, 3alpha,7beta,12alpha-Trihydroxy-5beta-cholanic acid, 3alpha,7beta,12alpha-Trihydroxy-5beta-cholanoic acid, 3alpha,7beta,12alpha-Trihydroxycholanic acid, 7-Epicholic acid, 7beta-Hydroxyisocholic acid, Ursocholic acid
IUPAC Name (4R)-4-[(3R,5S,7S,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Tekniske data

Quantity 10 mg
InChI Formula InChI=1S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29)/t13-,14+,15-,16-,17+,18+,19+,20+,22+,23+,24-/m1/s1
Formula Weight 408.2876
Percent Purity >95%
Chemical Name or Material Ursocholic Acid
Grade HPLC
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.