missing translation for 'onlineSavingsMsg'
Learn More

Ursocholic acid, MedChemExpress

Product Code. 16417497
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16417497 5 mg 5mg
16404768 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16417497 Supplier Medchem Express Supplier No. HY113212
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ursocholic acid, a bile acid found predominantly in bile of mammals, is transformed into deoxycholic acid by the intestinal microflora in mice. Ursodeoxycholic acid is an inhibitor of 7α-hydroxysteroid dehydrogenase and hepatocyte nuclear factor 1α.

Ursocholic acid, a bile acid found predominantly in bile of mammals, is transformed into deoxycholic acid by the intestinal microflora in mice. Ursodeoxycholic acid is an inhibitor of 7α-hydroxysteroid dehydrogenase and hepatocyte nuclear factor 1α.

Specifications

Chemical Name or Material Ursocholic acid
CAS 2955-27-3
Color White
Purity Grade Notes Research
Molecular Formula C24H40O5
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (244.76 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@H](CCC(O)=O)[C@H]1CC[C@@]2([H])[C@]3([H])[C@@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C
Molecular Weight (g/mol) 408.57
Formula Weight 408.57
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.