missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Urotensin II Receptor Monoclonal Antibody (443707)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA562011

Product Code. 30534109

  • £374.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The Urotensin-II receptor has been reported to be expressed at high levels in the heart and pancreas and also in artery and smooth muscle. Low transcript levels have been identified in brain, particularly in thalamus, and superior occipital gyrus. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Urotensin II Receptor
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q8VIH9
UTS2R
Mouse Urotensin-II R synthetic peptide.
100 μg
Primary
Mouse
Antibody
IgG2b
Flow Cytometry
443707
Unconjugated
UTS2R
G protein-coupled receptor 14; GPR14; G-protein coupled receptor 14; G-protein coupled sensory epithelial neuropeptide-like receptor; putative G protein-coupled receptor (SENR); Senr; Uiir; Uii-r; Uii-r1a; UII-receptor; UR2R; UR-2-R; UR-II-R; urotensin 2 receptor; urotensin II receptor; Urotensin-2 receptor; UT-II; UTIIR; UTR; UTR2; Uts2r
Rat
Protein A/G
RUO
217369
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato