missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urotensin-2 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33403-20ul
This item is not returnable.
View return policy
Description
Urotensin-2 Monoclonal antibody specifically detects Urotensin-2 in Rat samples. It is validated for ELISA,Western Blot
Specifications
| Urotensin-2 | |
| Monoclonal | |
| Western Blot 1:100 - 1:500, ELISA Recommended starting concentration is 1 μg/mL | |
| UCN2, UIIPRO1068, U-IIUrotensin II, urotensin 2, urotensin-2 | |
| A synthetic peptide corresponding to a sequence within amino acids 20-124 of human Urotensin-2 (UTS2). (NP_006777.1).,, Sequence:, SLPLLDSREISFQLSAPHEDARLTPEELERASLLQILPEMLGAERGDILRKADSSTNIFNPRGNLRKFQDFSGQDPNILLSHLLARIWKPYKKRETPDCFWKYCV | |
| 20 μL | |
| Cardiovascular Biology, Cell Biology, Signal Transduction | |
| 10911 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction