missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin III Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-33126
This item is not returnable.
View return policy
Description
Uroplakin III Polyclonal specifically detects Uroplakin III in Human samples. It is validated for Western Blot.
Specifications
| Uroplakin III | |
| Polyclonal | |
| Unconjugated | |
| 10 mM PBS with 0.05% Sodium Azide | |
| UPK3A | |
| A synthetic peptide, aa260-279 (GASESSYTSVNRGPPLDRAE) of human Uroplakin III (Uniprot: O75631) | |
| Protein A purified | |
| RUO | |
| Primary | |
| Uroplakin-3A (UPK3A) is a component of the asymmetric unit membrane (AUM). It is a highly specialized bio-membrane made by terminally differentiated urothelial cells. The protein may play an important role in AUM-cytoskeleton interaction in terminally differentiated urothelial cells. UPK3A also contributes to the formation of urothelial glycocalyx, which may play an important role in preventing bacterial adherence through FimH bacterial protein binding leading to bladder infection. UPK3A has been shown as a helpful marker for the detection of bladder cancer. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF | |
| 1.0 mg/mL | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, CyTOF-ready | |
| MGC119178, UP3a, UPIII, UPIIIA, UPK3, UPK3A, uroplakin 3, uroplakin 3A, Uroplakin III, uroplakin3a, uroplakin-3a | |
| Rabbit | |
| 30 kDa | |
| 0.1 mg | |
| Cancer | |
| 7380 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction