missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin III Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-88560
This item is not returnable.
View return policy
Description
Uroplakin III Polyclonal specifically detects Uroplakin III in Human samples. It is validated for Western Blot.
Specifications
| Uroplakin III | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| MGC119178, UP3a, UPIII, UPIIIA, UPK3, UPK3A, uroplakin 3, uroplakin 3A, Uroplakin III, uroplakin3a, uroplakin-3a | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of human Uroplakin III | |
| 100 μL | |
| Cancer | |
| 7380 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction