missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin II Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-33389
This item is not returnable.
View return policy
Description
Uroplakin II Polyclonal specifically detects Uroplakin II in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Uroplakin II | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| O00526 | |
| UPK2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ISSLSGLLSPALTESLLVALPPCHLTGGNATLMVRRANDSKVVTSSFVVPPCRGRRELVS | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| UP2, UPII, UPK2 uroplakin 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7379 | |
| Human | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion