missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin Ib, Mouse anti-Human, Clone: UPK1B/3081, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79799-100UG
This item is not returnable.
View return policy
Description
Uroplakin Ib Monoclonal specifically detects Uroplakin Ib in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Uroplakin Ib | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| tetraspanin-20, Tspan-20, TSPAN20tetraspan, UP1b, UPIb, UPK1, uroplakin 1B, Uroplakin Ib, uroplakin-1b | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 7348 | |
| Store at 4°C. | |
| IgG2b κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| UPK1B/3081 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| UPK1B | |
| Recombinant fragment (around aa 109-229) of human Uroplakin Ib protein (exact sequence is proprietary) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction