missing translation for 'onlineSavingsMsg'
Learn More

Uroplakin Ia, Mouse anti-Human, Clone: UPK1A/2925, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-79735-100UG

 View more versions of this product

Product Code. 18238137

  • £396.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Uroplakin Ia Monoclonal specifically detects Uroplakin Ia in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Uroplakin Ia
Monoclonal
0.2 mg/mL
ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA
Mouse
Protein A or G purified
RUO
11045
Store at 4°C.
IgG1 κ
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
UPK1A/2925
Unconjugated
10 mM PBS with 0.05% BSA
UPK1A
Recombinant fragment (around aa 114-173) of human Uroplakin Ia protein (exact sequence is proprietary)
100 μg
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.