missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin Ia, Mouse anti-Human, Clone: UPK1A/2921, Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £396.00
Specifications
| Antigen | Uroplakin Ia |
|---|---|
| Clone | UPK1A/2921 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18282577
|
Novus Biologicals
NBP2-79733-20UG |
20 μg |
£221.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18284366
|
Novus Biologicals
NBP2-79733-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Uroplakin Ia Monoclonal specifically detects Uroplakin Ia in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Uroplakin Ia | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA | |
| UPK1A | |
| IgG2b κ | |
| Protein A or G purified |
| UPK1A/2921 | |
| ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 11045 | |
| Recombinant fragment (around aa 114-173) of human Uroplakin Ia protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title