missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin Ia Antibody (UPK1A/8704R) [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-21073JF525
This item is not returnable.
View return policy
Description
Uroplakin Ia Monoclonal antibody specifically detects Uroplakin Ia in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications
| Uroplakin Ia | |
| Monoclonal | |
| Janelia Fluor 525 | |
| TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA | |
| Recombinant fragment (around aa 100-200) of human Uroplakin Ia protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Signal Transduction | |
| 11045 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| UPK1A/8704R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction