missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Urolithin A
GREENER_CHOICE

Product Code. p-7161691
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18756986

Brand: Tocris Bioscience™ 6762/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PI 3-K/Akt/mTOR signaling pathway inhibitor; blocks phosphorylation of Akt and p70S6K in pancreatic ductal adenocarcinoma (PDAC)

Urolithin A is a PI 3-K/Akt/mTOR signaling pathway inhibitor; blocks phosphorylation of Akt and p70S6K in pancreatic ductal adenocarcinoma (PDAC). Inhibits proliferation and migration and promotes apoptosis of PDAC cell lines in vitro (IC50 values are 10.23 - 62.28 μM). Suppresses tumor growth and improves survival in mice bearing pancreatic tumor xenografts. Also blocks amyloid-β fibrillation in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,8-Dihydroxy-6H-dibenzo[b,d]pyran-6-one
CAS 1143-70-0
Quantity 10 mg
Molecular Formula C13H8O4
Formula Weight 228.2
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.