missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UROD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565062
This item is not returnable.
View return policy
Description
Immunogen sequence: GLDWTVAPKKA RECVGKTVTL QGNLDPCALY ASEEEIGQLV KQMLDDFGPH RYIANLGHGL YPDMDPEHVG AFVDAVHKHS RLLRQN Highest antigen sequence identity to the following orthologs - mouse 93%, rat 94%.
This gene encodes the fifth enzyme of the heme biosynthetic pathway. This enzyme is responsible for catalyzing the conversion of uroporphyrinogen to coproporphyrinogen through the removal of four carboxymethyl side chains. Mutations and deficiency in this enzyme are known to cause familial porphyria cutanea tarda and hepatoerythropoetic porphyria.
Specifications
| UROD | |
| Polyclonal | |
| Unconjugated | |
| Urod | |
| AI323803; PCT; porphyrinogen carboxylase; porphyrinogen carboxy-lyase; UPD; URO D; UROD; Uro-d; Uroporphyrinogen decarboxylase; uroporphyrinogen III decarboxylase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7389 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P06132 | |
| Urod | |
| Recombinant Human UROD. Recombinant protein control fragment (Product #RP-104608). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction