missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Urocortin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104471
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Urocortin.
Urocortin (UCN), a 40 amino acids neuropeptide widely distributed in the central nervous system (CNS), belongs to a family of structurally related peptides including corticotrophin releasing factor (CRF), sauvagine and urotensin I. Urocortin is derived from a larger precursor protein (pro-urocortin, also named corticotensin). Urocortin has biological actions similar to CRF such as stress-related endocrine, autonomic and behavioral responses in mammalian species. It is a potent stimulator of adrenocorticotropic hormone (ACTH) secretion by corticotroph cells from the anterior pituitary in vitro and in vivo.
Specifications
| Urocortin | |
| Polyclonal | |
| Unconjugated | |
| Ucn | |
| Corticotensin; Mpv17; prepro-urocortin; Ucn; Ucn1; UI; UROC; Urocortin; urocortin, preproprotein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22226, 29151, 7349 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P55089, P55090, P81615 | |
| Ucn | |
| A synthesized peptide derived from human UCN(Accession P55089), corresponding to amino acid residues L19-G69. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction