missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Urine System Accessories

Product Code. 15412520
Change view
Click to view available options
Product Type:
Urine Transfer Device
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15412520 Urine Transfer Device Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15412520 Supplier Greiner Bio-One Supplier No. 450251
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

For hygienic transfer of urine from collection devices

TRUSTED_SUSTAINABILITY

Specifications

Product Type Urine Transfer Device
Material PET
Certifications/Compliance CE
For Use With (Application) For hygienic transfer of Urine from collection devices
Includes 10 cm Straw
Storage Requirements 4°C to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.