missing translation for 'onlineSavingsMsg'
Learn More

Uridine, TRC

Product Code. 30134059
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134059 5 g 5g
17930075 250 mg 250mg
17979334 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134059 Supplier Toronto Research Chemicals Supplier No. TRCU8299105G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-96-8
Molecular Formula C9 H12 N2 O6
Molecular Weight (g/mol) 244.2
Synonym 1-beta-D-Ribofuranosylpyrimidine-2,4(1H,3H)-dione, Adenosine Imp. F (EP), Uridine, Uracil, 1-β-D-ribofuranosyl- (7CI), 1-β-D-Ribofuranosyl-2,4(1H,3H)-pyrimidinedione, 1-β-D-Ribofuranosyluracil, NSC 20256, Uridin, β-D-Ribofuranoside, 2,4(1H,3H)-pyrimidinedione-1, β-Uridine
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC(=O)NC2=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15H,3H2,(H,10,13,16)/t4-,6-,7-,8-/m1/s1
Formula Weight 244.0695
Chemical Name or Material Uridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.