missing translation for 'onlineSavingsMsg'
Learn More

Uridine Phosphorylase 1/UPP1 Antibody (2F5), Novus Biologicals™

Product Code. 18364378 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364378 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364378 Supplier Novus Biologicals Supplier No. H00007378M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Uridine Phosphorylase 1/UPP1 Monoclonal antibody specifically detects Uridine Phosphorylase 1/UPP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Uridine Phosphorylase 1/UPP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2F5
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003355
Gene Alias EC 2.4.2.3, UDRPASE, UPASE, UPP, UPUPase 1, UrdPase 1, uridine phosphorylase, uridine phosphorylase 1
Host Species Mouse
Immunogen UPP1 (NP_003355, 1 a.a. ~ 92 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAATGANAEKAESHNDCPVRLLNPNIAKMKEDILYHFNLTTSRHNFPALFGDVKFVCVGGSPSRMKAFIRCVGAELGLDCPGRDYPNICAGT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7378
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.