missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Uridine, 99%

Product Code. 10091993 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10091993 1 g Glass bottle 1g
10274900 25 g Glass bottle 25g
10513691 100 g Glass bottle 100g
10012783 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10091993 Supplier Thermo Scientific Chemicals Supplier No. 140770010
  • ONLINE EXCLUSIVE PRICE
    £40.50
    £36.65 / 1g
    Save £3.85
    9% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58-96-8 | C9H12N2O6 | 244.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-96-8
Molecular Formula C9H12N2O6
Molecular Weight (g/mol) 244.2
MDL Number MFCD00006526
InChI Key DRTQHJPVMGBUCF-DHRBYNEYSA-N
Synonym uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd
PubChem CID 89495556
IUPAC Name 1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O

Specifications

Absorbance 0.72 to 0.76 (A250/A260), 0.34 to 0.38 (A280/A260)
Melting Point 162.0°C to 170.0°C
Color White
Loss on Drying 1% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 31, 23
Merck Index 15, 10062
Specific Rotation + 8.40
Spectrophotometry 98% min. (lambda max. 262nm,pH 7) molar absorptivity 10100+/-2%,on dry basis
Transmittance 95% min. (c=1, H2O, 430nm) 1cm cell
Formula Weight 244.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Uridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.