missing translation for 'onlineSavingsMsg'
Learn More

Uridine 5'-monophosphate, disodium salt, hydrate, 98%

Product Code. 15760839 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15760839 25 g Glass bottle 25g
10052890 1 g Glass bottle 1g
10762651 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15760839 Supplier Thermo Scientific Acros Supplier No. 226290250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

homogeneous | CAS: 3387-36-8 | C9H11N2Na2O9P | 368.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3387-36-8
Molecular Formula C9H11N2Na2O9P
Molecular Weight (g/mol) 368.15
MDL Number MFCD00149422
InChI Key KURVIXMFFSNONZ-IJRYJGAHNA-L
Synonym 5'-uridylicaciddisodiumsalt, uridine 5'-monophosphate, disodium salt, uridine-5'-monophosphate disodium salt
PubChem CID 123134060
SMILES [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=CC(=O)NC1=O

Specifications

Absorbance 0.71 to 0.77 (A250/A260),0.38 to 0.44 (A280/A260)
Melting Point 208.0°C to 210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 10, 9689
Solubility Information Solubility aq. soln.: ± 40% (20°C)
Spectrophotometry 97.5% min. (lambda max. 262 nm,pH 7) molar absorptivity 10000 ±2%,on dry basis
IUPAC Name disodium [(2R,3S,4S,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Transmittance 95% min. (c=1, 430 nm) 1 cm cell
Formula Weight 368.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Uridine 5 '-monophosphate, disodium salt, hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.