missing translation for 'onlineSavingsMsg'
Learn More

Uridine 5'-monophosphate, disodium salt, hydrate, 98%

Product Code. 10762651 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10762651

Brand: Thermo Scientific Acros 226290050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

homogeneous | CAS: 3387-36-8 | C9H11N2Na2O9P | 368.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3387-36-8
Molecular Formula C9H11N2Na2O9P
Molecular Weight (g/mol) 368.15
MDL Number MFCD00149422
InChI Key KURVIXMFFSNONZ-IJRYJGAHNA-L
Synonym uridine-5'-monophosphate disodium salt, uridine 5'-monophosphate, disodium salt, 5'-uridylicaciddisodiumsalt
PubChem CID 123134060
SMILES [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=CC(=O)NC1=O

Specifications

Absorbance 0.71 to 0.77 (A250/A260),0.38 to 0.44 (A280/A260)
Melting Point 208.0°C to 210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 10, 9689
Solubility Information Solubility in water:+/- 40% (20°C)
Spectrophotometry 97.5% min. (lambda max. 262nm,pH 7) molar absorptivity 10000+/-2%,on dry basis
IUPAC Name [(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
Transmittance 95% min. (c=1, 430nm) 1cm cell
Formula Weight 368.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Uridine 5′-monophosphate, disodium salt, hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.