missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Uridine 5'-monophosphate

Product Code. 16414756
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16414756 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16414756 Supplier MedChemExpress Supplier No. HY101981
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Uridine 5'-monophosphate (5'-​Uridylic acid), a monophosphate form of UTP, can be acquired either from a de novo pathway or degradation products of nucleotides and nucleic acids in vivo and is a major nucleotide analogue in mammalian milk.

Specifications

Molecular Weight (g/mol) 324.18 g/mol
Product Type Natural Products
Content And Storage Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Purity 97.0%
Quantity 100 mg
Solubility Information DMSO : 150 mg/mL (462.71 mM∣Need ultrasonic) ∣H2O : 150 mg/mL (462.71 mM∣Need ultrasonic)
For Use With (Application) Metabolism-protein/nucleotide metabolism
CAS 58-97-9
Synonym 5'-​Uridylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.