missing translation for 'onlineSavingsMsg'
Learn More

Uridine 5'-Diphosphoglucose Disodium Salt, TRC

Product Code. 30132419
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132419 500 mg 500mg
30137628 1 g 1g
30004764 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132419 Supplier Toronto Research Chemicals Supplier No. TRCU829950500MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28053-08-9
Molecular Formula C15H22N2Na2O17P2
Molecular Weight (g/mol) 610.27
Synonym Uridine 5'-(Trihydrogen diphosphate) P'-α-D-Glucopyranosyl Ester Disodium Salt, Uridine 5'-(Trihydrogen pyrophosphate) Mono-α-D-glucopyranosyl Ester Disodium Salt, Disodium UDP-glucose, Disodium Uridine Diphosphoglucose;
IUPAC Name disodium;[(2R,3S,4R,5R)-3,4-dihydroxy-5-(4-oxido-2-oxopyrimidin-1-yl)oxolan-2-yl]methyl [hydroxy-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphosphoryl] phosphate
SMILES C1=CN([C@H]2[C@@H]([C@@H]([C@@H](COP(=O)([O-])OP(=O)(O)O[C@@H]3[C@@H]([C@H]([C@@H]([C@@H](CO)O3)O)O)O)O2)O)O)C(=O)N=C1[O-].[Na+].[Na+]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H24N2O17P2.2Na/c18-3-5-8(20)10(22)12(24)14(32-5)33-36(28,29)34-35(26,27)30-4-6-9(21)11(23)13(31-6)17-2-1-7(19)16-15(17)25;;/h1-2,5-6,8-14,18,20-24H,3-4H2,(H,26,27)(H,28,29)(H,16,19,25);;/q;2*+1/p-2/t5-,6-,8-,9-,10+,11-,12-,13-,14-;;/m1../s1
Formula Weight 610.0189
Percent Purity >95%
Chemical Name or Material Uridine 5'-Diphosphoglucose Disodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.