Learn More
Uridine-5'-diphosphate trisodium salt, 98+%, Thermo Scientific Chemicals
Description
It has been used as a purinergic agonist to study its effects on absorptive cationic flux in cochlear outer sulcus cells (OSC) and vestibular transitional cells (VTC). Also been used for nucleoside diphosphatase (NDPase) activity assays in rabbit retinae.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It has been used as a purinergic agonist to study its effects on absorptive cationic flux in cochlear outer sulcus cells (OSC) and vestibular transitional cells (VTC). Also been used for nucleoside diphosphatase (NDPase) activity assays in rabbit retinae.
Solubility
Soluble in water to 100mM
Notes
Store in freezer -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Spezifikation
Spezifikation
| Color | White |
| Quantity | 100 mg |
| Solubility Information | Soluble in water to 100mM |
| Formula Weight | 470.11 |
| Percent Purity | ≥98% |
| Physical Form | Powder |
| Chemical Name or Material | Uridine-5'-diphosphate trisodium salt |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.