missing translation for 'onlineSavingsMsg'
Learn More

Uridine 5’-Diphosphate Sodium Salt, TRC

Product Code. 30449349
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449349 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449349 Supplier Toronto Research Chemicals Supplier No. TRCU82992050MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Uridine 5'-Diphosphate Sodium Salt
CAS 21931-53-3
Molecular Formula C9 H12 N2 O12 P2 . 2 Na
Quantity 50 mg
Synonym Uridine 5'-Diphosphate Sodium Salt
InChI Formula InChI=1S/C9H14N2O12P2.2Na/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(22-8)3-21-25(19,20)23-24(16,17)18;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,10,12,15)(H2,16,17,18);;/q;2*+1/p-2/t4-,6-,7-,8-;;/m1../s1
SMILES [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)([O-])OP(=O)(O)[O-])N2C=CC(=O)NC2=O
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate
Molecular Weight (g/mol) 448.12
Formula Weight 447.9661
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.