missing translation for 'onlineSavingsMsg'
Learn More

Uric acid, MedChemExpress

Product Code. 16422379
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422379 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422379 Supplier Medchem Express Supplier No. HYB2130
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Uric acid, scavenger of oxygen radical, is a very important antioxidant that help maintains the stability of blood pressure and antioxidant stress. Uric acid can remove reactive oxygen species (ROS) such as singlet oxygen and peroxynitrite, inhibiting lipid peroxidation.

Uric acid, scavenger of oxygen radical, is a very important antioxidant that help maintains the stability of blood pressure and antioxidant stress. Uric acid can remove reactive oxygen species (ROS) such as singlet oxygen and peroxynitrite, inhibiting lipid peroxidation.

Specifications

Chemical Name or Material Uric acid
CAS 69-93-2
Color White
Purity Grade Notes Research
Molecular Formula C5H4N4O3
Quantity 1 g
Solubility Information H2O : 10 mg/mL (59.48 mM; ultrasonic and adjust pH to 12 with NaOH) ∣DMSO : 1 mg/mL (5.95 mM; Need ultrasonic)
SMILES O=C(N1)NC2=C(NC(N2)=O)C1=O
Molecular Weight (g/mol) 168.11
Formula Weight 168.11
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.