missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RMP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567193
This item is not returnable.
View return policy
Description
Immunogen sequence: ILEEEPQENQK KLLPLSVTPE AFSGTVIEKE FVSPSLTPPP AIAHPALPTI PERKEVLLEA SEETGK Highest antigen sequence identity to the following orthologs - mouse 66%, rat 70%.
Involved in gene transcription regulation. Acts as a transcriptional repressor in concert with the corepressor UXT to regulate androgen receptor (AR) transcription. May act as a tumor suppressor to repress AR-mediated gene transcription and to inhibit anchorage-independent growth in prostate cancer cells. Required for cell survival in ovarian cancer cells. Together with UXT, associates with chromatin to the NKX3-1 promoter region. Antagonizes transcriptional modulation via hepatitis B virus X protein. Plays a central role in maintaining S6K1 signaling and BAD phosphorylation under normal growth conditions thereby protecting cells from potential deleterious effects of sustained S6K1 signaling. The URI1-PPP1CC complex acts as a central component of a negative feedback mechanism that counteracts excessive S6K1 survival signaling to BAD in response to growth factors. Mediates inhibition of PPP1CC phosphatase activity in mitochondria. Coordinates the regulation of nutrient-sensitive gene expression availability in a mTOR-dependent manner. Seems to be a scaffolding protein able to assemble a prefoldin-like complex that contains PFDs and proteins with roles in transcription and ubiquitination. [Uniprot].
Specifications
| RMP | |
| Polyclonal | |
| Unconjugated | |
| Uri1 | |
| C19orf2; C80913; FLJ10575; Nnx3; Ppp1r19; Protein NNX3; Protein phosphatase 1 regulatory subunit 19; protein phosphatase 1, regulatory subunit 19; RMP; RNA polymerase II subunit 5-mediating protein; RPB5-mediating protein; Unconventional prefoldin RPB5 interactor; unconventional prefoldin RPB5 interactor 1; URI; Uri1; URI1 prefoldin-like chaperone; URI1, prefoldin like chaperone; URI1, prefoldin-like chaperone | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8725 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94763 | |
| Uri1 | |
| Recombinant Human RMP. Recombinant protein control fragment (Product #RP-107832). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction