missing translation for 'onlineSavingsMsg'
Learn More

Urethane, MedChemExpress

Product Code. 16452319
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452319 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452319 Supplier Medchem Express Supplier No. HYB1207
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Urethane (Ethyl carbamate), the ethyl ester of carbamic acid, is a byproduct of fermentation found in various food products. Urethane has the ability to suppress bacterial, protozoal, sea urchin egg, and plant tissue growth in vitro.

Urethane (Ethyl carbamate), the ethyl ester of carbamic acid, is a byproduct of fermentation found in various food products. Urethane has the ability to suppress bacterial, protozoal, sea urchin egg, and plant tissue growth in vitro.

Specifications

Chemical Name or Material Urethane
CAS 51-79-6
Color White
Purity Grade Notes Research
Molecular Formula C3H7NO2
Synonym Ethyl carbamate Carbamic acid ethyl ester Ethylurethane
Solubility Information DMSO : 100 mg/mL (1122.46 mM; Need ultrasonic)
SMILES NC(OCC)=O
Molecular Weight (g/mol) 89.09
Quantity 1 g
Formula Weight 89.09
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.