missing translation for 'onlineSavingsMsg'
Learn More

Urethane-13C,15N, TRC

Product Code. 30455828
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455828 50 mg 50mg
30455586 10 mg 10mg
30453112 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455828 Supplier Toronto Research Chemicals Supplier No. TRCU82530750MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Urethane-13C,15N
Molecular Formula 13C C2 H7 15N O2
Quantity 50 mg
Synonym Carbamic Acid Ethyl Ester-13C,15N, Ethyl Aminoformate-13C,15N, Ethyl Carbamate-13C,15N, Ethylurethane-13C,15N, Leucethane-13C,15N, NSC 746-13C,15N, O-Ethylurethane-13C,15N, Pracarbamine-13C,15N, Urethan-13C,15N
InChI Formula InChI=1S/C3H7NO2/c1-2-6-3(4)5/h2H2,1H3,(H2,4,5)/i3+1,4+1
SMILES CCO[13C](=O)[15NH2]
IUPAC Name ethyl carbamate
Molecular Weight (g/mol) 91.079
Formula Weight 91.048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.