missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urethan, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Ethyl carbamate |
| CAS | 51-79-6 |
| Analyte or Component Names | Ethyl carbamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C3 H7 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ethyl aminoformate, Ethyl carbamate, Ethylurethane, Leucethane, NSC 746, O-Ethylurethane, Pracarbamine, Urethan, Urethane |
| InChI Formula | InChI=1S/C3H7NO2/c1-2-6-3(4)5/h2H2,1H3,(H2,4,5) |
| Solution Type | Neat |
| SMILES | CCOC(=O)N |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction