missing translation for 'onlineSavingsMsg'
Learn More

Urease, Jack Beans, Thermo Scientific™

Product Code. 15454049
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 15454049

Brand: Thermo Scientific Alfa Aesar J61455.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Suitable for the determination of urea. It is also used as chemical reagents, pharmaceutical intermediates. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-13-5
MDL Number MFCD00070858
Synonym EC 3.5.1.5,Urea amidohydrolase

Specifications

Packaging Plastic bottle
Quantity 1 g
Solubility Information It is slightly soluble in water.
Physical Form Lyophilized Powder
Chemical Name or Material Urease
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.