Learn More
Invitrogen™ Ureaplasma parvum (serovars 1, 3, 6, 14) Monoclonal Antibody (6527)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547570
Description
Specificity: Specific to the multi-banded antigen of U. parvum serovars. Reacts with serovars 1, 3, 6 & 14. These antibodies do not cross react with Mycoplasma hominis and Mycoplasma genitallium. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Specifications
| Ureaplasma parvum (serovars 1, 3, 6, 14) | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 6527 | |
| Unconjugated | |
| U. parvum | |
| Recombinant multi-banded antigen (MB) from Ureaplasma urealyticum, serovar 3. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.