missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea, U.S.P., J.T. Baker™
229.90 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Packaging | Poly Bottle |
Quantity | 500 g |
Specific Gravity | 1L = 1.32kg |
Formula Weight | 60.06 |
Grade | USP |
Chemical Name or Material | Urea |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'