missing translation for 'onlineSavingsMsg'
Learn More

Urea, MP Biomedicals

Product Code. 15709160 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
1 kg
25 lb.
Unit Size:
1kg
25lb
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15709160 1 kg 1kg
11404976 50 g 50g
11414976 25 lb. 25lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15709160 Supplier MP Biomedicals Supplier No. 04821528
  • ONLINE EXCLUSIVE PRICE
    £95.20
    £89.25 / 1kg
    Save £5.95
    6% Off
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • Dissolve urea in deionized water to the desired concentration
  • For every 10 ml of solution, add 1 g of Amberlite™ IRA-910
  • Stir for one hour at room temperature

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 130°C to 140°C
Color White
Boiling Point 135°C (decomposition)
Linear Formula CH4N2O
Quantity 1 kg
Solubility Information One gram dissolves in 1 mL water, 10 mL 95% ethanol, 1 mL boiling 95% ethanol, 20 mL absolute ethanol, 6 mL methanol, 2 mL glycerol; Soluble in concentrated hydrochloric acid; almost insoluble in chloroform, ether.
Formula Weight 60.06
Percent Purity ≥99%
Grade Ultra Pure
Physical Form Prills
Chemical Name or Material UREA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.