missing translation for 'onlineSavingsMsg'
Learn More

Urea, MedChemExpress

Product Code. 16439416
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16439416

Brand: Medchem Express HYY0271

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Urea is a powerful protein denaturant via both direct and indirect mechanisms. A potent emollient and keratolytic agent. Used as a diuretic agent. Blood urea nitrogen (BUN) has been utilized to evaluate renal function. Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry.

Urea is a powerful protein denaturant via both direct and indirect mechanisms. A potent emollient and keratolytic agent. Used as a diuretic agent. Blood urea nitrogen (BUN) has been utilized to evaluate renal function. Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry.

Specifications

Chemical Name or Material Urea
CAS 57-13-6
Color White
Purity Grade Notes Research
Molecular Formula CH4N2O
Quantity 100 mg
Synonym Carbamide Carbonyldiamide
Solubility Information H2O : 100 mg/mL (1665.00 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (1665.00 mM; Need ultrasonic)
SMILES NC(N)=O
Molecular Weight (g/mol) 60.06
Formula Weight 60.06
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.