missing translation for 'onlineSavingsMsg'
Learn More

Urea hydrogen peroxide adduct, 97%

Product Code. 11388835
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388835 500 g 500g
11368835 25 g 25g
11378835 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388835 Supplier Thermo Scientific Alfa Aesar Supplier No. L13940.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 124-43-6 | CH6N2O3 | 94.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Urea hydrogen peroxide is an antiseptic, disinfectant and bleaching agent used in pharmaceuticals and cosmetics respectively. It is used in the whitening of teeth and relieves minor inflammation of gums, oral mucosal surfaces and lips. It finds application in the preparation of plastics, blue print development and starch modification. In addition to this, it is used as a source of hydrogen peroxide easily handled in the laboratory.

Solubility
Soluble in water.

Notes
Incompatible with acetic anhydride, aniline and carboxylic acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-43-6
Molecular Formula CH6N2O3
Molecular Weight (g/mol) 94.07
MDL Number MFCD00013119
InChI Key AQLJVWUFPCUVLO-UHFFFAOYSA-N
Synonym urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid
PubChem CID 31294
ChEBI CHEBI:75178
IUPAC Name hydrogen peroxide;urea
SMILES C(=O)(N)N.OO

Specifications

Melting Point ∽90°C (decomposition)
Odor Odorless
Linear Formula H2NCONH2·H2O2
Quantity 500 g
UN Number UN1511
Beilstein 3680414
Sensitivity Very Hygroscopic
Merck Index 14,1782
Solubility Information Soluble in water.
Formula Weight 94.07
Percent Purity 97%
Chemical Name or Material Urea hydrogen peroxide adduct
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.